Craig Recrystallization Tube, DWK LIFE SCIENCES
Supplier: DWK Life Sciences (KIMBLE)
283550-0007
KT283550-0007EA 72.14 USD
KT283550-0007
Craig Recrystallization Tube, DWK LIFE SCIENCES
Tubes Recrystallization Tubes
A combination crystallizing and filtration centrifuge tube. Filtered solutions are allowed to cool in the Craig tube, inducing crystallization.
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • To balance in a centrifuge, two tubes are required
The Craig stopper is placed in the tube neck and the whole apparatus is inverted into a test or centrifuge tube.
Centrifuging forms a mat of crystals between the stopper and the walls of the Craig tube and draws away excess solvent. Further washing and crystallization steps are possible.
Ø×H: 14×100 mm
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.