Distillation Adapters, 105° Bend, with Vacuum take-off and Outer Hose Connection, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
276405-0000
KT276405-0000EA 114.53 USD
KT276405-0000
Distillation Adapters, 105° Bend, with Vacuum take-off and Outer Hose Connection, DWK Lifesciences
Distillation Components Distillation Adapters
These extended 105° bent distillation adapters with vacuum take-off and outer hose connection.
  • Barbed hose connection
  • Joints are the same size
Standard Taper inner joint at the bottom and Standard Taper outer joint at the top.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.