KIMBLE® KONTES® 105° Vacuum Take-Off Adapters, Standard Stem, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
205500-2440 276400-0000 276410-0000
KT205500-2440EA 167.06 USD
KT205500-2440 KT276400-0000 KT276410-0000
KIMBLE® KONTES® 105° Vacuum Take-Off Adapters, Standard Stem, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Adapters are bent at a 105° angle for use in vacuum distillation.
  • 105° angle bend
  • Top [ST] outer joint
  • Bottom [ST] inner joint
  • With hose connection and drip tip
Featuring [ST] top outer and lower inner joints with a hose connector and drip tip.