KIMBLE® Bleed-Type Inlet Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
275501-0000
KT275501-0000EA 50.32 USD
KT275501-0000
KIMBLE® Bleed-Type Inlet Tubes, DWK Life Sciences
Tubes Gas Inlet Tubes
This inlet tube is designed for the introduction of gases or liquids during reactions.
  • Hose connection fits ¼" Int.Ø flexible tubing
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Fits PTFE or glass universal adapters.