KIMBLE® KONTES® Inlet Adapter, Bleed-Type, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
273410-0000
KT273410-0000EA 22.92 USD
KT273410-0000
KIMBLE® KONTES® Inlet Adapter, Bleed-Type, DWK Life Sciences
Adapters Adapters, Conical Ground Joints
Liquids or gases may be directed into organic reactions using this adapter.
  • Bleed-type inlet adapter
  • Multiple uses
Adapter may also be calibrated and used as a pipet.