Distillation Adapters, 105° Bend, with Drip Tip, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
273200-0000
KT273200-0000EA 118.35 USD
KT273200-0000 KT273200-1922
Distillation Adapters, 105° Bend, with Drip Tip, DWK Life Sciences
Distillation Components Distillation Adapters
Bent 105° distillation adapters.
  • Standard taper inner drip joint at the bottom and standard taper outer joint at the top
  • Joints are the same size
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.