KIMBLE® Right Angle Flow Control Inlet Adapter with Glass Stopcock and Inner Standard Taper Joint, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
185000-2440
KT185000-2440EA 188.73 USD
KT185000-2440
KIMBLE® Right Angle Flow Control Inlet Adapter with Glass Stopcock and Inner Standard Taper Joint, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Use this adapter for regulating the input of gases or liquids into reaction systems.
  • Stopcock with glass plug
  • Inner [ST] joint on one end
  • 90° angle hose connection on opposite end
Adapter features an inner Standard Taper joint on one end and a 90° hose connection on the opposite end
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Stopcock has a glass plug.