KIMBLE® Bleed Type Inlet Adapter with Dual Hose Connections and Standard Taper Joint, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
181000-2440
KT181000-2440EA 128.92 USD
KT181000-2440
KIMBLE® Bleed Type Inlet Adapter with Dual Hose Connections and Standard Taper Joint, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Designed with two side hose connectors for venting or introducing gas/vacuum into a system.
  • Two side hose connectors
  • [ST] joint
  • Manufactured from borosilicate glass
The hose connections fit 3/8” ID flexible tubing.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.