KIMBLE® KONTES® Universal Inlet with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
179850-2114 179850-2124 179850-2214 179850-2224 179850-2324
KT179850-2114EA 185.83 USD
KT179850-2114 KT179850-2124 KT179850-2214 KT179850-2224 KT179850-2324
KIMBLE® KONTES® Universal Inlet with Hose Connection, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Designed to adapt a variety of tubes, condensers and filters in 14/20 and 24/25–24/40 glassware.
  • Inert O-ring
  • Vacuum tight seal
  • Serrated side port
  • Bottom inner joint
Diameter variations of 1 to 2mm can be accommodated, as the inert O-ring is compressed by the open-top polypropylene cap to effect a vacuum tight seal. The serrated side port can be used for a vacuum connection or for the introduction of gases. Bottom has an inner joint. Body is PTFE, O-rings are Viton, polypropylene compression cap is suitable for use to 150ºC, and side port is CTFE.
Ordering information: Includes Compression Cap.