KIMBLE® PTFE BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
179800-2124 179800-2214 179800-2224 179800-2324
KT179800-2124EA 149.19 USD
KT179800-2124 KT179800-2214 KT179800-2224 KT179800-2324
KIMBLE® PTFE BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
This is an adjustable fitting designed to adapt to a variety of apparatus.
  • Vacuum-tight seal
  • Bottom [ST] inner joint
  • Design allows use with clamps
  • FKM O-Rings and polypropylene cap included
A diameter variation of 1mm to 2mm can be accommodated, as the inert O-Ring is compressed by the open-top cap to create a vacuum-tight seal. The adapter body is made of PTFE. Bottom has a Standard Taper inner joint. Design allows use with clamps. Supplied with FKM O-Rings and with polypropylene cap suitable for use to 150°C (302°F).