KIMBLE® BEVEL-SEAL™ Thermometer Inlet Adapters with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
179750-0514 179750-0524 179750-0814 179750-1119 179750-2129 746035-0814 179750-2124
KT179750-0514CS 98.73 USD
KT179750-0514 KT179750-0524 KT179750-0814 KT179750-0824 KT179750-0829 KT179750-1119 KT179750-1129 KT179750-2129 KT746035-0510 KT746035-0814 KT179750-2114 KT179750-2124
KIMBLE® BEVEL-SEAL™ Thermometer Inlet Adapters with Hose Connection, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Designed for use with plain thermometers, pipets, syringes, or other small items with an O.D. from 2mm to 11mm.
  • Vacuum-tight seal
  • With hose connection
  • One FKM O-Ring and one cap included
Equipped with a hose connection for vacuum or the introduction of gases
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
A vacuum-tight seal is created with the open-top compression cap and FKM O-Ring to allow adjustable immersion of thermometers, etc. Cap is suitable for use to 150°C (302°F). To use as a septum port, replace the O-Ring with a PTFE-lined septum. Supplied with one FKM O-Ring and one cap.