Vacuum Filtration Adapters with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
178110-2440
KT178110-2440EA 182.76 USD
KT178110-2440
Vacuum Filtration Adapters with Hose Connection, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Vacuum filtration adapters with hose connection is used for reduced pressure filtration using glass vessel with standard taper joint.
  • Designed for pluro stopper
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Pluro stopper and funnel sold separately.