KIMBLE® Claisen Adapters for Distillation with BEVELSEAL™ Thermometer Joint, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
161100-2440
KT161100-2440CS 177.72 USD
KT161100-2440
KIMBLE® Claisen Adapters for Distillation with BEVELSEAL™ Thermometer Joint, DWK Life Sciences
Distillation Components Distillation Adapters
Used in distillation assemblies where adjustable immersion depth of the thermometer is desired.
  • Accommodation range 5 to 8 mm
  • Vacuum tight seal due to FKM O ring 010 size and open top compression cap
  • Threaded top BEVEL-SEAL™ connection for plain stem thermometers
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: KT410119-1307 is suitable for use to 200 °C.