KIMBLE® Enlarging Connecting Adapters, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
150100-0254 150100-0277 274750-0242 274750-0241 274750-0254
46200-578EA 97.23 USD
46200-578 46200-582 KT150100-0277 KT274750-0242 KT150100-0243 KT274750-0241 KT274750-0254
KIMBLE® Enlarging Connecting Adapters, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Enlarging Connecting Adapters are designed for enlarging the size of Standard Taper glass joints.
  • Top outer joint
  • Bottom inner joint
  • Manufactured from borosilicate glass
An outer joint is at the top, and an inner joint is at the bottom.
Adapters are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements.