KIMBLE® KONTES® Reducing Adapters, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
274750-0253 150100-0265 150100-0268 150100-0274 150100-0432 274750-0272 274750-0273
KT150100-0264EA 98.93 USD
KT150100-0264 KT274750-0253 KT150100-0261 KT150100-0265 KT150100-0268 KT150100-0274 KT150100-0432 KT274750-0263 KT274750-0272 KT274750-0273
KIMBLE® KONTES® Reducing Adapters, DWK Life Sciences
Adapters Adapters, Conical Ground Joints
Reducing adapters are used for connecting dissimilar [ST] joints.
  • Top outer joint
  • Bottom inner joint
The outer joint is at the top and inner joint is at the bottom.