Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

V643-07
JTV643-7CS 9860.97 USD
JTV643-7
Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
MW: 91.15 g/mol
Boiling Pt: 110 °C (1013 hPa)
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay ((CH₃)₄NOH) ≥9.8 %
Residue after Ignition ≤0.020 %
Ammonia and Other Amines Passes Test
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR