Tetrabutylammonium hydroxide 0.4 M in water for HPLC, for ion pair chromatography, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

V365-07
JTV365-7EA 672.35 USD
JTV365-7
Tetrabutylammonium hydroxide 0.4 M in water for HPLC, for ion pair chromatography, J.T.Baker®
Tetrabutylammonium hydroxide

Titrant, for use in ion-pair chromatography.

Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Molarity 0.37 - 0.43
Appearance Passes Test
Absorbance at 420 nm (neat) ≤0.150
Bromide (Br) ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR