4-Aminobutyric acid ≥99%, Biotech Reagent

Supplier: Avantor


Management of Change (MOC) category = R

V265-10
JTV265-10EA 90840.5 USD
JTV265-10
4-Aminobutyric acid ≥99%, Biotech Reagent
4-Aminobutyric acid
Formula: C₄H₉NO₂
Melting Pt: 195…204 °C
MDL Number: MFCD00008226
CAS Number: 56-12-2

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (White to off-white solid) Passes Test
Identification (FTIR) Passes Test
Clarity of solution (5% in water) Passes Test
Assay ≥99 %
Loss on drying (105°C) ≤0.5 %
Residual solvent - Methanol ≤3000 ppm
Residual solvent - Ethanol ≤5000 ppm
Residual solvent - Isopropanol ≤5000 ppm
Aluminum (Al) ≤60 ppm
Cadmium (Cd) ≤2 ppm
Chromium (Cr) ≤7 ppm
Cobalt (Co) ≤100 ppm
Copper (Cu) ≤20 ppm
Manganese (Mn) ≤10 ppm
Zinc (Zn) ≤900 ppm
Nickel (Ni) ≤20 ppm
Selenium (Se) ≤25 ppm
Bacterial endotoxins, EU/g (For information only)
Total Aerobic Microbial Count, For Information Only
Total Yeasts and Molds Count, For Information Only
Residual Solvents - Only Class 3 solvents are likely to be present from
the product purification process. Class 3 solvents are identified,
quantified and reported for each production batch.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR