Propylene carbonate, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester

Management of Change (MOC) category = N

U497-09
JTU497-9CS 1247.19 USD
JTU497-9
Propylene carbonate, J.T.Baker®
Propylene carbonate
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 241…242 °C (1013 hPa)
Melting Pt: –48.8 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 132 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR