2-Phenoxyethanol

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

T319-07 T319-09
JTT319-7CS 1304.38 USD
JTT319-7 JTT319-9
2-Phenoxyethanol
2-Phenoxyethanol
Formula: C₆H₅OCH₂CH₂OH
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 13 °C
Density: 1.1094 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002857
CAS Number: 122-99-6
EINECS: 204-589-7
Merck Index: 12,07410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27cc4d39b806528798164961c23a1285d41d6a9&vid=a46b721cc69101d0c50dc49d9687445b395a3766&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR