Sodium hypochlorite (5% Cl₂) in aqueous solution, Laboratory Reagent, sterile, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

P005-03
JTP005-3CS 924.81 USD
JTP005-3
Sodium hypochlorite (5% Cl₂) in aqueous solution, Laboratory Reagent, sterile, J.T.Baker®
Sodium hypochlorite

BAKER - A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).

Formula: ClNaO
MW: 74.44 g/mol
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –30…–20 °C
Density: 1.22…1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,II

Order Now

Specification Test Results

CAUTION: For Manufacturing, processing or repackaging
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Assay (NaClO) 4.0 - 6.0 %
Mercury (Hg) ≤5.0 ppb
The above referenced lot has passed the current USP Test for Sterility.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR