Ethanol denatured solution 70%, sterile made with water for injection (WFI) (denatured with methanol and isopropanol), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Ethyl alcohol denatured

Management of Change (MOC) category = NI

P004-03 BAKRP004-03
JTP004-3CS 1065.39 USD
JTP004-3 SCJTP004-3
Ethanol denatured solution 70%, sterile made with water for injection (WFI) (denatured with methanol and isopropanol), J.T.Baker®
Ethanol denatured
Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Specific Gravity 0.882 - 0.896
Nonvolatile Residue Passes Test
Assay 68.0 - 72.0 %
Irradiation: Minimum Delivered Dose (K Gy) 25.0 - 50.0
Irradiation: Maximum Delivered Dose (K Gy) 25.0 - 50.0
Irradiation: Specified Dose Range (Lower) 25.0 - 50.0
Irradiation: Specified Dose Range (Upper) 25.0 - 50.0
Irradiation Date
Irradiation Batch Number
The above referenced lot has passed the current USP Test for Sterility.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR