2-Propanol solution ≥70%, sterile, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

P002-03
JTP002-3CS 1061.25 USD
JTP002-3
2-Propanol solution ≥70%, sterile, J.T.Baker®
2-Propanol

Made with USP Water for Injection

Formula: (CH₃)₂CHOH
Density: 0.8…0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1987
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

CAUTION: For Manufacturing, processing or repackaging
USP - Specific Gravity at 20°/20°C 0.872 - 0.883
Appearance Passes Test
USP - Acidity (mL) ≤1.0
USP - Nonvolatile Residue (mg) ≤5.0
USP - Assay 68.0 - 72.0 %
Irradiation: Minimum Delivered Dose (K Gy) 25.0 - 50.0
Irradiation: Maximum Delivered Dose (K Gy) 25.0 - 50.0
Irradiation: Specified Dose Range (Lower) 25.0 - 50.0
Irradiation: Specified Dose Range (Upper) 25.0 - 50.0
Irradiation Date
Irradiation Batch Number
The above referenced lot has passed the current USP Test for Sterility.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR