Imidazole 99,0, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

M.O.C. Category NI

N811-06 N811-05 N811-07
JTN811-6EA 686.69 USD
JTN811-6 JTN811-5 JTN811-07
Imidazole 99,0, BAKER ANALYZED® ACS
Imidazole
Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8 III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₃H₄N₂) ≥99.0 %
Trace Impurities - Iron (Fe) ≤0.001 %
pH of 5% Solution at 25°C 9.5 - 11.0
Residue after Ignition ≤0.10 %
Water (H₂O) ≤0.2 %
DNase Activity, For Information Only Passes Test
Protease Activity, For Information Only Passes Test
RNase Activity, For Information Only Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR