Hexane (mixture of isomers) ≥95% (by GC, sum of isomers + methylcyclopentane), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hexanes, Isohexane

Management of Change (MOC) category = N

N169-08
JTN169-8CS 610.85 USD
JTN169-8
Hexane (mixture of isomers) ≥95% (by GC, sum of isomers + methylcyclopentane), J.T.Baker®
Hexane (mixture of isomers)

BAKER - A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (sum of 5 isomers, total hexanes, plus methylcyclopentane)(by GC) ≥95 %
Identification (by IR) Passes Test
Appearance (Clear, colorless liquid) Passes Test
Color (APHA) ≤10
Residue after Evaporation ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR