1,1,1,3,3,3-Hexafluoro-2-propanol 99 GC, corrected for water content, BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

M.O.C. Category N

N151-05
JTN151-5EA 1234.88 USD
JTN151-5
1,1,1,3,3,3-Hexafluoro-2-propanol 99 GC, corrected for water content, BAKER ANALYZED®
1,1,1,3,3,3-Hexafluoro-2-propanol

Lot analysis on label.

Formula: C₃H₂F₆O
MW: 168.04 g/mol
Boiling Pt: 59.1
Melting Pt: -96
Density: 1.492 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 1760
ADR: 8 II
Merck Index: 13,09598

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₃H₂F₆0) (by GC, corrected for water) ≥99 %
Color (APHA) ≤10
Residue after Evaporation ≤0.100 %
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR