L(+)-Glutamic acid ≥99.0% (dried basis, T), BAKER ANALYZED®, Biochemical Reagent

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

M756-07
JTM756-7EA 256.22 USD
JTM756-7
L(+)-Glutamic acid ≥99.0% (dried basis, T), BAKER ANALYZED®, Biochemical Reagent
L(+)-Glutamic acid

Lot analysis on label.

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.538 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₅H₉NO₄) (dried basis, by acid-base titrn) ≥99.0 %
Specific Rotation, [Å]²º^D(dried basis, c=2 in 5N HCl)(+) 29.9 - 32.1 Degree
Homogeneity by TLC Passes Test
Ash (sulfated) ≤0.1 %
Loss on Drying at 85°C ≤0.1 %
Chloride (Cl) ≤0.2 %
Arsenic (As) ≤0.0003 %
Heavy Metals (as Pb) ≤0.002 %
Iron (Fe) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR