Giemsa's stain stock solution 8.0 g/L in glycerol/methanol (1:1) for hematology, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
Management of Change (MOC) category = NI
M708-01
JTM708-1CS
3177.39
USD
JTM708-1
Giemsa's stain stock solution 8.0 g/L in glycerol/methanol (1:1) for hematology, J.T.Baker®
Giemsa's stain
For staining blood cells. Contains 8.0 g/L giemsa stain, 500 ml/L anhydrous glycerol and 500 ml/L absolute methanol. Prepared with certified stain.
Density:
0.99 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00081642 CAS Number: 51811-82-6 EINECS: 257-438-2 UN: 1230 ADR: 3,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Suitability for Blood Cell Staining | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...