Formamide ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Carbamaldehyde

Management of Change (MOC) category = N

M522-06
JTM522-6CS 6160.79 USD
JTM522-6
Formamide ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Formamide

Lot analysis on label.

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HCONH₂) (by GC) ≥99.5 %
Freezing Point 2.0 - 3.0 °C
Color (APHA) ≤10
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR