Iron(III) citrate monohydrate, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

M376-07
JTM376-7CS 3558.39 USD
JTM376-7
Iron(III) citrate monohydrate, J.T.Baker®
Iron (III) citrate monohydrate
Formula: C₆H₅FeO₇·H₂O
MDL Number: MFCD00013098
CAS Number: 2338-05-8
EINECS: 249-117-0

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as Fe) ≥16.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR