Diethylene glycol monoethyl ether, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

L216-07
JTL216-7EA 130.1 USD
JTL216-7
Diethylene glycol monoethyl ether, J.T.Baker®
Diethylene glycol monoethyl ether
Formula: C₆H₁₄O₃
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: –80 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 13,01809

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Identification (by IR) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR