Calcium sulfate, Drierite™ (10-20 Mesh), with indicator (cobalt (II) chloride), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

L059-07
JTL059-7CS 3218.73 USD
JTL059-7
Calcium sulfate, Drierite™ (10-20 Mesh), with indicator (cobalt (II) chloride), J.T.Baker®
Calcium sulphate
Formula: CaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR