Calcium sulfate, Drierite™ (4 Mesh), with indicator (cobalt (II) chloride), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite

L057-02 L057-07
JTL057-2CS 2283.25 USD
JTL057-2 JTL057-7
Calcium sulfate, Drierite™ (4 Mesh), with indicator (cobalt (II) chloride), J.T.Baker®
Calcium sulphate
Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1350
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
UN: 3077
Merck Index: 13,01711

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR