Sodium dodecyl sulfate (SDS) ≥95%, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
L050-07
L050-02
JTL050-7CS
572.5
USD
JTL050-7
JTL050-02
Sodium dodecyl sulfate (SDS) ≥95%, J.T.Baker®
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 204…207 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 2926 ADR: 4.1, 6.1,II Merck Index: 14,08636 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (as C₁₂H₂₅OSO₃Na) | ≥95 % |
Identification (by IR) | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...