Dithiothreitol (DTT, Cleland's reagent) ≥99.0%, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

Management of Change (MOC) category = R

F781-05
JTF781-5EA 25703.39 USD
JTF781-5
Dithiothreitol (DTT, Cleland's reagent) ≥99.0%, Biotech Reagent, J.T.Baker®
Dithiothreitol (DTT, Cleland's reagent)

White crystals. Keep material refrigerated between 2 to 8°C.

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: ∼125 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Keep refrigerated between 2-8°C (36-46°F) and store under nitrogen.
CAUTION: For Manufacturing, processing or repackaging
Assay (HSCH₂(CHOH)₂CH₂SH) ≥99.0 %
Appearance Passes Test
Absorbance of a 0.1M Solution (1-cm path vs water) at - 280 nm ≤0.060
Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm ≤0.400
Infrared Spectrum Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR