Carbon disulfide ≥99.9% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for spectroscopy, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
E350-01
JTE350-1CS
3176.85
USD
JTE350-1
Carbon disulfide ≥99.9% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for spectroscopy, J.T.Baker®
Carbon disulphide
Suitable for NIOSH analytical methods 1003, 1500, and 1501 for hydrocarbon analysis and spectroscopy.
Formula:
CS₂ Boiling Pt: 46.2 °C (1013 hPa) Melting Pt: –111.6 °C Density: 1.2632 g/cm³ (20 °C) Flash Pt: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011321 CAS Number: 75-15-0 EINECS: 200-843-6 UN: 1131 ADR: 3 (6.1),I Merck Index: 13,01821 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (CS₂) (by GC, corrected for water) | ≥99.9 % |
ACS - Trace Organic Impurities (as benzene) (by GC) | ≤1 ppm |
ACS - Color (APHA) | ≤10 |
ACS - Residue after Evaporation | ≤15 ppm |
ACS - Hydrogen Sulfide (H₂S) | ≤1.5 ppm |
ACS - Sulfur Dioxide (SO₂) | ≤2.5 ppm |
ACS - Water (H₂O)(by Karl Fischer titrn) | ≤0.05 % |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 750 nm | ≤0.01 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 500 nm | ≤0.01 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 410 nm | ≤0.05 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 403 nm | ≤0.10 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 394 nm | ≤0.25 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 388 nm | ≤0.50 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 385 nm | ≤1.00 |
Boiling Point (typical)................. 46.3 °C. | |
Windows of Near Infrared Transmittance (10-mm path vs. air 80-90% T), | |
µm: | |
2.68 - 3.34; 2.23 - 2.68; 0.75 - 2.19 | |
Windows of Infrared Transmittance (10-mm path vs air, 60-100% T), µm: | |
2.5 - 4.3; 4.8 - 6.2 ; 7.1 - 16.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...