1-Butanesulfonic acid sodium salt ≥98%, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butanesulfonate, Butane-1-sulfonic acid sodium salt, Sodium 1-butansulphonate, Sodium 1-butansulfonate, Butane-1-sulphonic acid sodium salt, Sodium butane-1-sulfonate
Management of Change (MOC) category = N
D600-03
JTD600-3EA
530.13
USD
JTD600-3
1-Butanesulfonic acid sodium salt ≥98%, J.T.Baker®
1-Butansulphonic acid sodium salt
Powder.
Formula:
C₄H₉NaO₃S MW: 160.17 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007540 CAS Number: 2386-54-1 EINECS: 219-201-1 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (C₄H₉NaO₃) (by titration) | ≥98 % |
Appearance | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...