Hydrochloric acid concentrate for dilution to 0.1 mol (0.1N), DILUT-IT® dissolution media concentrate, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

D011-00
JTD011-0CS 699.46 USD
JTD011-0
Hydrochloric acid concentrate for dilution to 0.1 mol (0.1N), DILUT-IT® dissolution media concentrate, J.T.Baker®
Hydrochloric acid

Clear, colorless solution. Dissolution media concentrates are produced from raw materials in accordance with USP, Ph. Eur., BP, and JP guidelines under validated processes in Mallinckrodt Baker's ISO certified manufacturing facilities. Infinitely soluble.

Formula: HCl
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
STORE AT ROOM TEMPERATURE
Appearance Passes Test
Density (g/mL) at 25°C, For Information Only
Normality at 26x dilution 0.098 - 0.102
DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR