Potassium dihydrogen phosphate GMP Manufactured Product

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

CH10-60
JTCH10-60EA 7110.64 USD
JTCH10-60
Potassium dihydrogen phosphate GMP Manufactured Product
Potassium dihydrogen phosphate
Formula: KH₂PO₄
MDL Number: MFCD00011401
CAS Number: 7778-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR