Liquid Level Indicators, J.T.Baker®, Avantor
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
9803-03
JT9803-03CS 1358.16 USD
JT9803-03
Liquid Level Indicators, J.T.Baker®, Avantor
Level Sensors
These level indicators provide the digital readout of the approximate volume of solvent remaining in the CYCLE-TAINER system.
  • Signal can also be used to drive switching circuits