Ammonia solution 30%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

9733-03 9733-01
JT9733-3CS 1069.94 USD
JT9733-3 JT9733-1
Ammonia solution 30%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Ammonia solution

For trace metal analysis. Lot analysis on label.

Formula: NH₃
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.88…0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as NH₃) 28.0 - 30.0 %
Specific Gravity at 60°/60°F 0.896 - 0.902
Appearance Passes Test
Color (APHA) ≤10
Residue after Ignition ≤3 ppm
Substances Reducing Permanganate Passes Test
Chloride (Cl) ≤0.5 ppm
Phosphate (PO₄) ≤0.4 ppm
Total Sulfur (as SO₄) ≤1 ppm
Trace Impurities - Aluminum (Al) ≤20 ppb
Trace Impurities - Arsenic and Antimony (as As) ≤50.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Boron (B) ≤20.0 ppb
Trace Impurities - Cadmium (Cd) ≤5.0 ppb
Trace Impurities - Calcium (Ca) ≤200.0 ppb
Trace Impurities - Chromium (Cr) ≤5.0 ppb
Trace Impurities - Cobalt (Co) ≤1.0 ppb
Trace Impurities - Copper (Cu) ≤20.0 ppb
Heavy Metals (as Pb) ≤200.0 ppb
Trace Impurities - Iron (Fe) ≤10.0 ppb
Trace Impurities - Lead (Pb) ≤5.0 ppb
Trace Impurities - Lithium (Li) ≤20.0 ppb
Trace Impurities - Magnesium (Mg) ≤20 ppb
Trace Impurities - Manganese (Mn) ≤5.0 ppb
Trace Impurities - Mercury (Hg) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤5.0 ppb
Trace Impurities - Potassium (K) ≤500.0 ppb
Trace Impurities - Silicon (Si) ≤1000.0 ppb
Trace Impurities - Silver (Ag) ≤5.0 ppb
Trace Impurities - Sodium (Na) ≤300 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤5.0 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR