Ammonia solution 28,0 - 30,0% (NH₃ basis), BAKER ANALYZED® A.C.S. Reagent ACS analytical reagent
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
Management of Change (MOC) category = NI
9721-03
9721-05
9721-02
9721-06
9721-00
9721-01
9721-15
9721-33
BAKR9721-00
JT9721-3CS
466.04
USD
JT9721-3
JT9721-5
JT9721-2
JT9721-6
JT9721-0
JT9721-1
JT9721-15
JT9721-33
SCJT9721-0
SCJT9721-1
SCJT9721-33
JT9721-18
Ammonia solution 28,0 - 30,0% (NH₃ basis), BAKER ANALYZED® A.C.S. Reagent ACS analytical reagent
Ammonia solution
The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.
Formula:
NH₃ MW: 35.05 g/mol Boiling Pt: 32…37.7 °C (1013 hPa) Melting Pt: –60…–57.5 °C Density: 0.88…0.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8 III |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For laboratory, research or manufacturing use. | |
Appearance (colorless and free from suspended matter or sediment) | Passes Test |
Assay (as NH₃) | 28.0 - 30.0% |
Color (APHA) | ≤5 |
Specific Gravity at 60/60 °F | 0.896 - 0.902 |
Residue after Ignition | ≤0.0020% |
Carbon Dioxide (CO₂) | ≤0.002% |
Substances Reducing Permanganate | Passes Test |
Chloride (Cl) | ≤0.5 ppm |
Nitrate (NO₃) | ≤2 ppm |
Phosphate (PO₄) | ≤2 ppm |
Sulfate (SO₄) | ≤2 ppm |
Trace Impurities - Aluminum (Al) | ≤200.0 ppb |
Arsenic and Antimony (as As) | ≤3000.0 ppb |
Trace Impurities - Barium (Ba) | ≤300.0 ppb |
Trace Impurities - Boron (B) | ≤50 ppb |
Trace Impurities - Chromium (Cr) | ≤100.0 ppb |
Trace Impurities - Copper (Cu) | ≤100.0 ppb |
Trace Impurities - Gold (Au) | ≤200.0 ppb |
Heavy Metals (as Pb) | ≤500.0 ppb |
Trace Impurities - Iron (Fe) | ≤100.0 ppb |
Trace Impurities - Lead (Pb) | ≤200.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤200.0 ppb |
Trace Impurities - Manganese (Mn) | ≤100.0 ppb |
Trace Impurities - Nickel (Ni) | ≤100.0 ppb |
Trace Impurities - Tin (Sn) | ≤100 ppb |
Trace Impurities - Titanium (Ti) | ≤100.0 ppb |
Trace Impurities - Zinc (Zn) | ≤100.0 ppb |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...