Nitric acid 65.0 - 71.0%, CMOS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Nitric acid solution

Management of Change (MOC) category = R

9685-69
JT9685-69EA 3.77 USD
JT9685-69
Nitric acid 65.0 - 71.0%, CMOS, J.T.Baker®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

For Microelectronic Use
Assay (HNO₃) 65.0 - 71.0 %
Color, APHA ≤10
Chloride (Cl) ≤80 ppb
Sulfate (SO4) ≤500 ppb
Phosphate (PO₄) ≤200 ppb
Arsenic (As) ≤100.0 ppb
Copper (Cu) ≤100.0 ppb
Lead (Pb) ≤100.0 ppb
Iron (Fe) ≤100.0 ppb
Sodium (Na) ≤100.0 ppb
Boron (B) ≤100.0 ppb
Magnesium (Mg) ≤100.0 ppb
Aluminum (Al) ≤100.0 ppb
Potassium (K) ≤100.0 ppb
Calcium (Ca) ≤100.0 ppb
Chromium (Cr) ≤100.0 ppb
Nickel (Ni) ≤100.0 ppb
Zinc (Zn) ≤100.0 ppb
Silver (Ag) ≤100.0 ppb
Barium (Ba) ≤100.0 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR