Nitric acid fuming ≥90.0% (by titrimetric analysis), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Nitric acid solution

Management of Change (MOC) category = N

9624-02 9624-05
JT9624-2EA 494.06 USD
JT9624-2 JT9624-5
Nitric acid fuming ≥90.0% (by titrimetric analysis), BAKER ANALYZED® ACS, J.T.Baker®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,I
REACH: 01-2119487297-23
Merck Index: 13,06610

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (as HNO₃) (by acidimetry) ≥90.0 %
Dilution Test Passes Test
ACS - Residue after Ignition ≤0.002 %
ACS - Dissolved Oxides (as N₂O₃) ≤0.1 %
ACS - Chloride (Cl) ≤0.7 ppm
ACS - Sulfate (SO₄) ≤5 ppm
Trace Impurities - Arsenic (As) ≤0.3 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR