Nitric acid 69,0 - 70,0%, CMOS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Nitric acid solution

Management of Change (MOC) category = R

9606-03
JT9606-33CS 388.43 USD
JT9606-33 JT9606-3
Nitric acid 69,0 - 70,0%, CMOS, J.T.Baker®
Nitric acid
Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

Assay (HNO₃} 69.0 - 70.0 %
Appearance Passes Test Passes Test
Color (APHA) ≤10
Residue after Ignition ≤2 ppm
Chloride (Cl) ≤0.08 ppm
Phosphate (PO₄) ≤0.1 ppm
Sulfate (SO₄) ≤0.2 ppm
Trace Impurities - Aluminum (Al) ≤40.0 ppb
Arsenic and Antimony (as As) ≤5 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤20.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤50.0 ppb
Trace Impurities - Calcium (Ca) ≤50.0 ppb
Trace Impurities - Chromium (Cr) ≤30.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤20.0 ppb
Trace Impurities - Gold (Au) ≤20.0 ppb
Heavy Metals (as Pb) ≤100 ppb
Trace Impurities - Iron (Fe) ≤40.0 ppb
Trace Impurities - Lead (Pb) ≤20.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤20.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Nickel (Ni)
Trace Impurities - Niobium (Nb)
Trace Impurities - Potassium (K) ≤50.0 ppb
Trace Impurities - Silicon (Si) ≤50.0 ppb
Trace Impurities - Silver (Ag) ≤20.0 ppb
Trace Impurities - Sodium (Na) ≤150.0 ppb
Trace Impurities - Strontium (Sr) ≤30.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤20.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤20.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
Particle Count - 0.5 µm and greater ≤60 par/ml
Particle Count - 1.0 µm and greater ≤10 par/ml
For Microelectronic Use
Storage Note: Do not store near Ammonium Hydroxide. White crystals may form around the opening. The crystals can be removed with deionized water. This will not affect the quality of the product in the bottle.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR