Nitric acid 69.0 - 70.0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

9598-34 9598-00 9598-05 9598-51
JT9598-34CS 718.82 USD
JT9598-34 JT9598-0 JT9598-5 JT9598-51
Nitric acid 69.0 - 70.0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Nitric acid

For trace metal analysis. Lot analysis on label.

Formula: HNO₃
Melting Pt: –42 °C
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (HNO₃) 69.0 - 70.0 %
Appearance Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Ignition ≤2 ppm
Chloride (Cl) ≤0.04 ppm
Phosphate (PO₄) ≤0.10 ppm
Sulfate (SO₄) ≤0.4 ppm
Trace Impurities - Aluminum (Al) ≤30.0 ppb
Arsenic and Antimony (as As) ≤5.0 ppb
Trace Impurities - Barium (Ba) ≤1.0 ppb
Trace Impurities - Beryllium (Be) ≤1.0 ppb
Trace Impurities - Bismuth (Bi) ≤1.0 ppb
Trace Impurities - Boron (B) ≤4.0 ppb
Trace Impurities - Cadmium (Cd) ≤1.0 ppb
Trace Impurities - Calcium (Ca) ≤50.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Cobalt (Co) ≤1.0 ppb
Trace Impurities - Copper (Cu) ≤1.0 ppb
Trace Impurities - Gallium (Ga) ≤20.0 ppb
Trace Impurities - Germanium (Ge) ≤4.0 ppb
Trace Impurities - Gold (Au) ≤4.0 ppb
Heavy Metals (as Pb) ≤100 ppb
Trace Impurities - Iron (Fe) ≤10.0 ppb
Trace Impurities - Lead (Pb) ≤0.5 ppb
Trace Impurities - Lithium (Li) ≤1.0 ppb
Trace Impurities - Magnesium (Mg) ≤7.0 ppb
Trace Impurities - Manganese (Mn) ≤1.0 ppb
Trace Impurities - Mercury (Hg) ≤0.5 ppb
Trace Impurities - Molybdenum (Mo) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤1.0 ppb
Trace Impurities - Niobium (Nb) ≤1.0 ppb
Trace Impurities - Potassium (K) ≤5.0 ppb
Trace Impurities - Silicon (Si) ≤20.0 ppb
Trace Impurities - Silver (Ag) ≤1.0 ppb
Trace Impurities - Sodium (Na) ≤200.0 ppb
Trace Impurities - Strontium (Sr) ≤1.0 ppb
Trace Impurities - Tantalum (Ta) ≤2.0 ppb
Trace Impurities - Thallium (Tl) ≤5.0 ppb
Trace Impurities - Tin (Sn) ≤5.0 ppb
Trace Impurities - Vanadium (V) ≤1.0 ppb
Trace Impurities - Zinc (Zn) ≤5.0 ppb
Trace Impurities - Zirconium (Zr) ≤1.0 ppb
Meets ACS Specifications


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR