Acetic acid glacial ≥99.7% for HPLC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9515-03
JT9515-3CS 989.04 USD
JT9515-3
Acetic acid glacial ≥99.7% for HPLC, J.T.Baker®
Acetic acid

BAKER ANALYZED™ HPLC Reagent—High purity solvents for use in analytical and preparative separations. All are optimized for all of your Liquid Chromatography applications, including HPLC, UHPLC, gel permeation chromatography and UV-spectrophotometric analysis.

Formula: H₃CCOOH
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
Assay (by GC, corrected for water) ≥99.7 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.01
Residue after Evaporation ≤0.0005 %
Water (by Karl Fischer titrn) ≤0.1 %
(1) Snyder, L.R., J. Chromatography, 92,223-230 (1974)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR