Toluene ≥99.5% (by GC), CMOS for microelectronic, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

Management of Change (MOC) category = R

9476-09
JT9476-9EA 1289.22 USD
JT9476-9
Toluene ≥99.5% (by GC), CMOS for microelectronic, J.T.Baker®
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Microelectronic Use
Assay (C₆H₅CH₃) (by GC) ≥99.5 %
Color (APHA) ≤10
Acidity (µeq/g) ≤0.2
Residue after Evaporation ≤2.0 ppm
Water (by KF, coulometric) ≤0.03 %
Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤0.003 %
Chloride (Cl) ≤2 ppm
Phosphate (PO₄) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤20 ppb
Arsenic and Antimony (as As) ≤10.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Boron (B) ≤20.0 ppb
Trace Impurities - Cadmium (Cd) ≤20.0 ppb
Trace Impurities - Calcium (Ca) ≤100.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Cobalt (Co) ≤20 ppb
Trace Impurities - Copper (Cu) ≤20.0 ppb
Trace Impurities - Gallium (Ga) ≤50 ppb
Trace Impurities - Germanium (Ge) ≤50.0 ppb
Trace Impurities - Gold (Au) ≤20 ppb
Heavy Metals (as Pb) ≤500.0 ppb
Trace Impurities - Iron (Fe) ≤20.0 ppb
Trace Impurities - Lithium (Li) ≤20.0 ppb
Trace Impurities - Magnesium (Mg) ≤10.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤20.0 ppb
Trace Impurities - Potassium (K) ≤50 ppb
Trace Impurities - Silicon (Si) ≤100.0 ppb
Trace Impurities - Silver (Ag) ≤20.0 ppb
Trace Impurities - Sodium (Na) ≤100.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤30.0 ppb
Trace Impurities - Zinc (Zn) ≤20.0 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR