Toluene ≥99.5%, extremely low particle, CMOS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = R
9466-03
JT9466-3CS
699.9
USD
JT9466-3
Toluene ≥99.5%, extremely low particle, CMOS, J.T.Baker®
Toluene
Formula:
C₆H₅CH₃ Boiling Pt: 110.6 °C (1013 hPa) Melting Pt: –95 °C Density: 0.867 g/cm³ (20 °C) Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II REACH: 05-2114615130-69 Merck Index: 13,09607 |
|
Specification Test Results
For Microelectronic Use | |
Assay (C₆H₅CH₃) (by GC) | ≥99.5 % |
Color (APHA) | ≤10 |
Acidity (µeq/g) | ≤0.2 |
Residue after Evaporation | ≤2.0 ppm |
Water (by KF, coulometric) | ≤0.03 % |
Substances Darkened by H₂SO₄ | Passes Test |
Sulfur Compounds (as S) | ≤0.003 % |
Chloride (Cl) | ≤2 ppm |
Phosphate (PO₄) | ≤0.5 ppm |
Trace Impurities - Aluminum (Al) | ≤20 ppb |
Arsenic and Antimony (as As) | ≤10.0 ppb |
Trace Impurities - Barium (Ba) | ≤10.0 ppb |
Trace Impurities - Boron (B) | ≤20.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤20.0 ppb |
Trace Impurities - Calcium (Ca) | ≤100.0 ppb |
Trace Impurities - Chromium (Cr) | ≤10.0 ppb |
Trace Impurities - Cobalt (Co) | ≤20 ppb |
Trace Impurities - Copper (Cu) | ≤20.0 ppb |
Trace Impurities - Gallium (Ga) | ≤50 ppb |
Trace Impurities - Germanium (Ge) | ≤50.0 ppb |
Trace Impurities - Gold (Au) | ≤20 ppb |
Heavy Metals (as Pb) | ≤500.0 ppb |
Trace Impurities - Iron (Fe) | ≤20.0 ppb |
Trace Impurities - Lithium (Li) | ≤20.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤10.0 ppb |
Trace Impurities - Manganese (Mn) | ≤10.0 ppb |
Trace Impurities - Nickel (Ni) | ≤20.0 ppb |
Trace Impurities - Potassium (K) | ≤50 ppb |
Trace Impurities - Silicon (Si) | ≤100.0 ppb |
Trace Impurities - Silver (Ag) | ≤20.0 ppb |
Trace Impurities - Sodium (Na) | ≤100.0 ppb |
Trace Impurities - Strontium (Sr) | ≤10.0 ppb |
Trace Impurities - Tin (Sn) | ≤30.0 ppb |
Trace Impurities - Zinc (Zn) | ≤20.0 ppb |
Particle Count - 1.0 µm and greater | ≤10 par/ml |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...