Tetrachloroethylene ≥99.0% (by GC) stabilized, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Stabilization: Stabilizer 3-5 ppm
Management of Change (MOC) category = N

9465-01 9465-03
JT9465-1CS 1734.12 USD
JT9465-1 JT9465-3
Tetrachloroethylene ≥99.0% (by GC) stabilized, BAKER ANALYZED®, J.T.Baker®
Tetrachloroethylene

Contains approximately 3 to 5 ppm stabilizer. Lot analysis on label.

Formula: C₂Cl₄
Boiling Pt: 121.1 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III
Merck Index: 12,09332
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Cl₂C:CCl₂) (by GC) ≥99.0 %
Residue after Evaporation ≤0.0050 %
Acidity (as HCl) ≤0.002 %
Alkalinity (as NaOH) ≤0.002 %
Free Chlorine (as Cl₂) Passes Test
Chloride (Cl) ≤1.0 ppm
Water (H₂O)(by Karl Fischer titrn) ≤0.005 %
Contains 3 - 5 ppm of stabilizer.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR